Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 2-cyanoacrylate

Catalog No. E15055G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E15055G 5 g
E150510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E15055G Supplier Sigma Aldrich Supplier No. E15055G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl 2-cyanoacrylate (ECA) is a monomeric adhesive that can be used on a variety of materials such as metals, plastics, rubber, ceramics, and wood. It forms a one part adhesive system that can also be used as a primer.ECA can be used as a tissue adhesive for potential applications in cardiovascular and pulmonary surgeries. It can also be mixed with hydrophobic silica nanoparticles for the designing of cotton fabrics for medical applications.

Specifications

CAS 7085-85-0
Molecular Formula C6H7NO2
Linear Formula C6H7NO2
Quantity 5 g
MDL Number MFCD00045615
Molecular Weight (g/mol) 125.13
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.