Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Cyanopropionate 97.0+%, TCI America™
SDP

Catalog No. C10835ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10835ML 5 mL
C108325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10835ML Supplier TCI America Supplier No. C10835ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1572-99-2
Molecular Formula C6H9NO2
Molecular Weight (g/mol) 127.143
MDL Number MFCD00013812
InChI Key MIHRVXYXORIINI-UHFFFAOYSA-N
Synonym 2-Cyanopropionic Acid Ethyl Ester
PubChem CID 98018
ChEBI CHEBI:51925
IUPAC Name ethyl 2-cyanopropanoate
SMILES CCOC(=O)C(C)C#N

Specifications

Color Yellow
Boiling Point 193°C
Quantity 5 mL
UN Number 3276
Formula Weight 127.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Cyanopropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.