Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 2-cyclohexanoneacetate

Catalog No. E1980425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1980425G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E1980425G Supplier Sigma Aldrich Supplier No. E1980425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl 2-cyclohexanoneacetate

Specifications

CAS 24731-17-7
Density 1.02 g/mL (at 25°C (literature))
Molecular Formula C10H16O3
Refractive Index n20/D 1.458 (literature)
Linear Formula (O=)C6H9CH2CO2C2H5
MDL Number MFCD00001638
Quantity 250 mg
Synonym Ethyl 2-oxocyclohexaneacetate
Molecular Weight (g/mol) 184.23
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.