Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 2-deoxy-2-phthalimido-β-D-thioglucopyranoside | 130539-43-4 | ≥98.0% | 353.39 g/mol | C16H19NO6S | 500 MG
SDP

Catalog No. 5000299823
Encompass_Preferred

Ethyl 2-deoxy-2-phthalimido-β-D-thioglucopyranoside is a specialized glycobiology reagent used in carbohydrate chemistry and oligosaccharide synthesis. It functions as an ethyl thio-glycoside donor and is supplied for laboratory research applications where glycosylation steps or mechanistic studies of carbohydrate reactions are required.

  • Used as a glycosyl donor in carbohydrate synthesis.
  • Suitable for glycobiology and carbohydrate chemistry research.
  • Molecular formula C16H19NO6S.
  • Molecular weight 353.39 g/mol.
  • Provided as a stable solid for laboratory-scale experiments.
  • Recommended storage maintains long-term stability under cooled conditions.

Catalog No. 50-002-99823 Supplier Medchemexpress LLC Supplier No. HYW039873500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 2-deoxy-2-phthalimido-β-D-thioglucopyranoside is a specialized glycobiology reagent used in carbohydrate chemistry and oligosaccharide synthesis. It functions as an ethyl thio-glycoside donor and is supplied for laboratory research applications where glycosylation steps or mechanistic studies of carbohydrate reactions are required.

  • Used as a glycosyl donor in carbohydrate synthesis.
  • Suitable for glycobiology and carbohydrate chemistry research.
  • Molecular formula C16H19NO6S.
  • Molecular weight 353.39 g/mol.
  • Provided as a stable solid for laboratory-scale experiments.
  • Recommended storage maintains long-term stability under cooled conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.