Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 2-deoxy-2-phthalimido-β-D-thioglucopyranoside | 130539-43-4 | MFCD28167068 | 97.0% | 353.39 g/mol | C16H19NO6S | 1 G
SDP

Catalog No. 5000301213
Encompass_Preferred

Ethyl 2-deoxy-2-phthalimido-β-D-thioglucopyranoside is a biochemical reagent used in glycobiology and carbohydrate chemistry research. Supplied as a powder, it is used in studies of glycan formation and degradation, glycosylation processes, and protein-glycan interactions in basic research, biomedicine, and biotechnology.

  • Molecular formula: C16H19NO6S
  • Molecular weight: 353.39 g/mol
  • Appearance: powder
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
  • Intended use: biochemical assays and glycobiology research

Catalog No. 50-003-01213 Supplier Medchemexpress LLC Supplier No. HYW0398731G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 2-deoxy-2-phthalimido-β-D-thioglucopyranoside is a biochemical reagent used in glycobiology and carbohydrate chemistry research. Supplied as a powder, it is used in studies of glycan formation and degradation, glycosylation processes, and protein-glycan interactions in basic research, biomedicine, and biotechnology.

  • Molecular formula: C16H19NO6S
  • Molecular weight: 353.39 g/mol
  • Appearance: powder
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
  • Intended use: biochemical assays and glycobiology research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.