Learn More
Medchemexpress LLC Ethyl 2-deoxy-2-phthalimido-β-D-thioglucopyranoside | 130539-43-4 | MFCD28167068 | 98.0% | 353.39 g/mol | C16H19NO6S | 10 G
SDP
Catalog No. 5000299452
Encompass_Preferred
Ethyl 2-deoxy-2-phthalimido-β-D-thioglucopyranoside is a protected thioglycoside reagent used in carbohydrate chemistry and glycosylation research. It is supplied as a powder and is characterized by CAS 130539-43-4, molecular formula C16H19NO6S, and molecular weight 353.39 g/mol.
  • Available as a powdered reagent suitable for laboratory use.
  • High reported purity (98.0%).
  • Stable under recommended storage conditions: powder -20°C (3 years) or 4°C (2 years).
  • In solvent storage recommendations: -80°C (6 months) or -20°C (1 month).
  • Applicable for carbohydrate synthesis and glycosylation studies.
  • Molecular formula C16H19NO6S and molecular weight 353.39 g/mol.
Catalog No. 50-002-99452 Supplier Medchemexpress LLC Supplier No. HYW03987310G
Ethyl 2-deoxy-2-phthalimido-β-D-thioglucopyranoside is a protected thioglycoside reagent used in carbohydrate chemistry and glycosylation research. It is supplied as a powder and is characterized by CAS 130539-43-4, molecular formula C16H19NO6S, and molecular weight 353.39 g/mol.
  • Available as a powdered reagent suitable for laboratory use.
  • High reported purity (98.0%).
  • Stable under recommended storage conditions: powder -20°C (3 years) or 4°C (2 years).
  • In solvent storage recommendations: -80°C (6 months) or -20°C (1 month).
  • Applicable for carbohydrate synthesis and glycosylation studies.
  • Molecular formula C16H19NO6S and molecular weight 353.39 g/mol.