Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 2-deoxy-2-phthalimido-β-D-thioglucopyranoside | 130539-43-4 | MFCD28167068 | 98.0% | 353.39 g/mol | C16H19NO6S | 250 MG
SDP

Catalog No. 5000303462
Encompass_Preferred

Ethyl 2-deoxy-2-phthalimido-β-D-thioglucopyranoside (CAS 130539-43-4) is a phthalimido-protected thio-β-D-glucopyranoside used as a research reagent in carbohydrate chemistry and synthetic method development. It features a phthalimido group at the 2-position and an ethyl thio linkage at the anomeric center; molecular formula C16H19NO6S, molecular weight 353.39 g/mol. For research use only.

  • Phthalimido protecting group at the 2-position.
  • Ethyl thio linkage at the anomeric center.
  • Molecular formula C16H19NO6S; molecular weight 353.39 g/mol.
  • Appearance: powder.
  • Suitable for carbohydrate synthesis and method development.
  • Typical purity reported as 98% (supplier dependent).
  • For research use only; not for human or clinical use.

Catalog No. 50-003-03462 Supplier Medchemexpress LLC Supplier No. 250MGHYW039873
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 2-deoxy-2-phthalimido-β-D-thioglucopyranoside (CAS 130539-43-4) is a phthalimido-protected thio-β-D-glucopyranoside used as a research reagent in carbohydrate chemistry and synthetic method development. It features a phthalimido group at the 2-position and an ethyl thio linkage at the anomeric center; molecular formula C16H19NO6S, molecular weight 353.39 g/mol. For research use only.

  • Phthalimido protecting group at the 2-position.
  • Ethyl thio linkage at the anomeric center.
  • Molecular formula C16H19NO6S; molecular weight 353.39 g/mol.
  • Appearance: powder.
  • Suitable for carbohydrate synthesis and method development.
  • Typical purity reported as 98% (supplier dependent).
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.