Learn More
Medchemexpress LLC Ethyl 2-(diethoxyphosphoryl)propanoate | 3699-66-9 | 99.35% | 238.22 | 25 G
SDP
Catalog No. 5000453253
Encompass_Preferred
Ethyl 2-(diethoxyphosphoryl)propanoate is a biochemical reagent for life science research. It functions as a Wittig-Horner reagent. It can be used as a biological material or organic compound, and it condenses with a wide variety of aldehydes and ketones to form acrylates. This product is for research use only and not sold to patients.
Catalog No. 50-004-53253 Supplier Medchemexpress LLC Supplier No. 25GHYW013436
Ethyl 2-(diethoxyphosphoryl)propanoate is a biochemical reagent for life science research. It functions as a Wittig-Horner reagent. It can be used as a biological material or organic compound, and it condenses with a wide variety of aldehydes and ketones to form acrylates. This product is for research use only and not sold to patients.