Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 2-ethoxy-2-iminoacetate

Catalog No. 790419500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-041-9500MG 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 79-041-9500MG Supplier Sigma Aldrich Supplier No. 790419500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl 2-ethoxy-2-iminoacetate

Specifications

CAS 816-27-3
Density 1.042 g/mL (at 25°C)
Molecular Formula C6H11NO3
Refractive Index n20/D 1.426
Linear Formula C6H11NO3
Quantity 10 g
Synonym (1-Ethoxyformimidoyl)formic acid ethyl ester; 2-Ethoxy-2-iminoacetic acid ethyl ester; Ethyl 1-carbethoxyformimidate; Ethyl carboethoxyformidimate; Ethyl ethoxycarbonylformimidate; Ethyl ethoxyiminoacetate
Molecular Weight (g/mol) 145.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.