Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aobchem Ethyl 2-ethylbenzoate | ≥95% | CAS 56427-44-2 | C11H14O2 | 10g

Catalog No. 5001061514
Encompass_Preferred

Ethyl 2-ethylbenzoate | ≥95% | CAS 56427-44-2 | C11H14O2 | 10g

Catalog No. 50-010-61514 Supplier Aobchem Supplier No. A7998610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 2-ethylbenzoate | ≥95% | CAS 56427-44-2 | C11H14O2 | 10g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.