Learn More
Medchemexpress LLC Ethyl 2-Hydroxybutyr 1g
SDP
Catalog No. 5000206279
Encompass_Preferred
Ethyl 2-Hydroxybutyrate has a sweet fruity odor and is commonly used as a flavoring agent in the food and beverage industry In addition it can be used as an intermediate in the synthesis of various organic compounds including fragrances pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial processes notably in the production of cosmetics personal care products and cleaning agents
Catalog No. 50-002-06279 Supplier Medchemexpress LLC Supplier No. HYW1275001G
Ethyl 2-Hydroxybutyrate has a sweet fruity odor and is commonly used as a flavoring agent in the food and beverage industry In addition it can be used as an intermediate in the synthesis of various organic compounds including fragrances pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial processes notably in the production of cosmetics personal care products and cleaning agents