Learn More
Medchemexpress LLC Ethyl 2-Hydroxybutyr 25g
SDP
Catalog No. 5000205459
Encompass_Preferred
Ethyl 2-Hydroxybutyrate has a sweet fruity odor and is commonly used as a flavoring agent in the food and beverage industry In addition it can be used as an intermediate in the synthesis of various organic compounds including fragrances pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial processes notably in the production of cosmetics personal care products and cleaning agents
Catalog No. 50-002-05459 Supplier Medchemexpress LLC Supplier No. 25GHYW127500
Ethyl 2-Hydroxybutyrate has a sweet fruity odor and is commonly used as a flavoring agent in the food and beverage industry In addition it can be used as an intermediate in the synthesis of various organic compounds including fragrances pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial processes notably in the production of cosmetics personal care products and cleaning agents