Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-(Hydroxymethyl)acrylate (stabilized with HQ) 95.0+%, TCI America™
SDP

Catalog No. H091625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H091625G 25 g
H09165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H091625G Supplier TCI America Supplier No. H091625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10029-04-6
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD01673859
InChI Key SYGAXBISYRORDR-UHFFFAOYSA-N
Synonym 2-(Hydroxymethyl)acrylic Acid Ethyl Ester
PubChem CID 24827
IUPAC Name ethyl 2-(hydroxymethyl)prop-2-enoate
SMILES CCOC(=O)C(=C)CO

Specifications

Color Yellow
Boiling Point 80°C
Quantity 25 g
UN Number 2810
Formula Weight 130.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-(Hydroxymethyl)acrylate (stabilized with HQ)
EINECSNumber (2)-3861
RTECSNumber AT1835000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.