Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Isopropylacetoacetate 94.0+%, TCI America™
SDP

Catalog No. E111925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E111925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E111925ML Supplier TCI America Supplier No. E111925ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1522-46-9
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.22
MDL Number MFCD00042884
InChI Key DMIFFKCVURTPTG-UHFFFAOYNA-N
Synonym ethyl 2-isopropylacetoacetate, 2-isopropylacetoacetic acid ethyl ester, ethyl 2-iso-propylacetoacetate, butanoic acid, 2-acetyl-3-methyl-, ethyl ester, ethyl alpha-isopropylacetoacetate, ethyl 2-acetyl-3-methylbutyrate, acetoacetic acid, 2-isopropyl-, ethyl ester, ethyl .alpha.-isopropylacetoacetate, ethyl isopropylaceto-acetate, acmc-1c35s
PubChem CID 95544
IUPAC Name ethyl 2-acetyl-3-methylbutanoate
SMILES CCOC(=O)C(C(C)C)C(C)=O

Specifications

Color Colorless
Boiling Point 202°C
Quantity 25 mL
Formula Weight 172.22
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Isopropylacetoacetate
EINECSNumber (2)-1505
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.