Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 2-isopropylthiazole-4-carboxylate

Catalog No. SYX003291G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX003291G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX003291G Supplier Sigma Aldrich Supplier No. SYX003291G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Hexanoyl chloride was used in:
  • Total synthesis of (+/-)-7-butyl-6,8-dihydroxy-3-pentyl-3,4-dihydroisochromen-1-one
  • Asymmetric synthesis of 14-methyl-1-octadecene, the sex pheromone of the peach leafminer moth
  • Preparation of hexanoyl-coated nanofibers dispersible in several organic solvents
  • Total synthesis of natural isocarbostyril ruprechstyril (3-n-pentyl-6-methoxy-8-hydroxy-1(2h)-isoquinolinone), isolated from ruprechtia tangarana
  • Synthesis of 5-chloro-8-hydroxy-6-methoxy-3-pentylisocoumarin, the 5-chloro analog of naturally occurring 7-chloro-8-hydroxy-6-methoxy-3-pentylisocoumarin, isolated from tessmannia densiflora

Specifications

CAS 133047-44-6
Molecular Formula C9H13NO2S
Linear Formula C9H13NO2S
MDL Number MFCD09025787
Quantity 100 mL
Molecular Weight (g/mol) 199.27
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.