Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-mercaptopropionate, 98%, Thermo Scientific™

Catalog No. AAL1152014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1152014 25 g
AAL1152006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1152014 Supplier Thermo Scientific Chemicals Supplier No. L1152014

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19788-49-9
Molecular Formula C5H10O2S
Molecular Weight (g/mol) 134.193
MDL Number MFCD00040233
InChI Key LXXNWCFBZHKFPT-UHFFFAOYSA-N
Synonym ethyl 2-mercaptopropionate, ethyl 2-mercaptopropanoate, ethyl thiolactate, propanoic acid, 2-mercapto-, ethyl ester, 2-mercaptopropionic acid ethyl ester, propionic acid, 2-mercapto-, ethyl ester, ethyl alpha-mercaptopropionate, fema no. 3279, ethyl2-mercaptopropionate, ethyl mercaptopropionate
PubChem CID 519709
IUPAC Name ethyl 2-sulfanylpropanoate
SMILES CCOC(=O)C(C)S

Specifications

Density 1.043
Boiling Point 75°C to 76°C (35 mmHg)
Flash Point 46°C (115°F)
Odor Strong
Refractive Index 1.45
Quantity 25 g
UN Number UN1993
Beilstein 1071465
Sensitivity Air Sensitive
Formula Weight 134.2
Percent Purity 98%
Chemical Name or Material Ethyl 2-mercaptopropionate
Show More Show Less
Hazard Category H226-H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 243-314-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.