Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Mercaptopropionate 97.0+%, TCI America™
SDP

Catalog No. M103725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M103725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M103725G Supplier TCI America Supplier No. M103725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19788-49-9
Molecular Formula C5H10O2S
Molecular Weight (g/mol) 134.193
MDL Number MFCD00040233
InChI Key LXXNWCFBZHKFPT-UHFFFAOYSA-N
Synonym ethyl 2-mercaptopropionate, ethyl 2-mercaptopropanoate, ethyl thiolactate, propanoic acid, 2-mercapto-, ethyl ester, 2-mercaptopropionic acid ethyl ester, propionic acid, 2-mercapto-, ethyl ester, ethyl alpha-mercaptopropionate, fema no. 3279, ethyl2-mercaptopropionate, ethyl mercaptopropionate
PubChem CID 519709
IUPAC Name ethyl 2-sulfanylpropanoate
SMILES CCOC(=O)C(C)S

Specifications

Color Colorless
Boiling Point 90°C
Quantity 25 g
UN Number 1993
Formula Weight 134.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Mercaptopropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.