Learn More
Medchemexpress LLC Ethyl 2-methylpentanoate | 39255-32-8 | MFCD00036564 | 99.8% | 144.21 g·mol-1 | C8H16O2 | 50 G
SDP
Catalog No. 5000300073
Encompass_Preferred
Ethyl 2-methylpentanoate is an organic ester (CAS 39255-32-8) used as a biochemical reagent and as an intermediate in flavor and fragrance applications. It is typically a colorless to pale yellow liquid with reported purity around 99.78%, molecular formula C8H16O2, and molecular weight 144.21 g·mol-1, supplied in laboratory pack sizes for research use.
  • Used as a flavor and fragrance intermediate and research reagent.
  • High purity suitable for analytical applications.
  • Colorless to pale yellow liquid at room temperature.
  • Molecular formula C8H16O2 and molecular weight 144.21 g·mol-1.
  • Available in small laboratory pack sizes for convenient handling.
Catalog No. 50-003-00073 Supplier Medchemexpress LLC Supplier No. HYW01566750G
Ethyl 2-methylpentanoate is an organic ester (CAS 39255-32-8) used as a biochemical reagent and as an intermediate in flavor and fragrance applications. It is typically a colorless to pale yellow liquid with reported purity around 99.78%, molecular formula C8H16O2, and molecular weight 144.21 g·mol-1, supplied in laboratory pack sizes for research use.
  • Used as a flavor and fragrance intermediate and research reagent.
  • High purity suitable for analytical applications.
  • Colorless to pale yellow liquid at room temperature.
  • Molecular formula C8H16O2 and molecular weight 144.21 g·mol-1.
  • Available in small laboratory pack sizes for convenient handling.