Learn More
Medchemexpress LLC Ethyl 2-methylpentanoate | 39255-32-8 | MFCD00036564 | 99.8% | 144.21 g/mol | C8H16O2 | 500 G
SDP
Catalog No. 5000300517
Encompass_Preferred
Ethyl 2-methylpentanoate is a colorless liquid ester used as a biochemical reagent, flavor and fragrance ingredient, solvent, and synthetic intermediate. It has formula C8H16O2, molecular weight 144.21 g/mol, density ≈0.864 g/cm3, and is typically supplied at high purity for research applications.
  • High purity (~99.8%) for consistent experimental results.
  • Colorless liquid form, easy to measure and dispense.
  • Useful as a flavoring or fragrance ingredient with a fruity odor.
  • Serves as a solvent or plasticizer in formulation applications.
  • Suitable as a starting material for organic synthesis.
Catalog No. 50-003-00517 Supplier Medchemexpress LLC Supplier No. HYW015667500G
Ethyl 2-methylpentanoate is a colorless liquid ester used as a biochemical reagent, flavor and fragrance ingredient, solvent, and synthetic intermediate. It has formula C8H16O2, molecular weight 144.21 g/mol, density ≈0.864 g/cm3, and is typically supplied at high purity for research applications.
  • High purity (~99.8%) for consistent experimental results.
  • Colorless liquid form, easy to measure and dispense.
  • Useful as a flavoring or fragrance ingredient with a fruity odor.
  • Serves as a solvent or plasticizer in formulation applications.
  • Suitable as a starting material for organic synthesis.