Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Nonynoate, TCI America™
SDP

Catalog No. N059625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N059625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N059625ML Supplier TCI America Supplier No. N059625ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10031-92-2
Molecular Formula C11H18O2
Molecular Weight (g/mol) 182.263
MDL Number MFCD00036553
InChI Key BFZNMUGAZYAMTG-UHFFFAOYSA-N
Synonym ethyl 2-nonynoate, 2-nonynoic acid, ethyl ester, ethyl-2-nonynoate, ethyl nonynoate, ethyl octynecarboxylate, 2-nonynoic acid ethyl ester, ethyl octinecarbonate, unii-853ded009t, fema no. 2448, nonynoic acid, ethyl ester
PubChem CID 61451
IUPAC Name ethyl non-2-ynoate
SMILES CCCCCCC#CC(=O)OCC

Specifications

Color Colorless
Boiling Point 125°C
Quantity 25 mL
Formula Weight 182.26
Physical Form Liquid
Chemical Name or Material Ethyl 2-Nonynoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.