Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-oxo-4-phenylbutyrate, 90+%

Catalog No. AAB2014809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2014809 10 g
AAB2014818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2014809 Supplier Thermo Scientific Chemicals Supplier No. B2014809
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 64920-29-2 | C12H14O3 | 206.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64920-29-2
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD00037533
InChI Key STPXIOGYOLJXMZ-UHFFFAOYSA-N
Synonym ethyl 2-oxo-4-phenylbutyrate, ethyl benzylpyruvate, ethyl-2-oxo-4-phenylbutyrate, unii-5ynu9lpt51, 5ynu9lpt51, 2-oxo-4-phenylbutyric acid ethyl ester, 2-oxo-4-phenyl butyric acid ethyl ester, ethyl 2-oxo-4-phenyl-butanoate, 2-oxo-4-phenylbutanoic acid ethyl ester, ethyl2-oxo-4-phenylbutyrate
PubChem CID 562087
IUPAC Name ethyl 2-oxo-4-phenylbutanoate
SMILES CCOC(=O)C(=O)CCC1=CC=CC=C1

Specifications

Density 1.09
Boiling Point 131°C to 133°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.502
Quantity 10 g
Beilstein 2725083
Formula Weight 206.24
Percent Purity ≥90%
Chemical Name or Material Ethyl 2-oxo-4-phenylbutyrate
EINECSNumber 265-276-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.