Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 2-(phenylcarbonothioylthio)-2-phenylacetate

Catalog No. 7735061G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-350-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 77-350-61G Supplier Sigma Aldrich Supplier No. 7735061G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl 2-(phenylcarbonothioylthio)-2-phenylacetate

Specifications

CAS 1150308-13-6
Melting Point 48°C to 53°C
Molecular Formula C17H16O2S2
Linear Formula C17H16O2S2
Quantity 1 g
MDL Number MFCD24386381
Synonym alpha-[(Phenylthioxomethyl)thio]benzeneacetic acid ethyl ester; dithiobenzoate RAFT; Benzodithioate phenyl acetic acid ethyl ester
Molecular Weight (g/mol) 316.44
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.