Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Undecynoate 97.0+%, TCI America™
SDP

Catalog No. U006425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U006425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U006425ML Supplier TCI America Supplier No. U006425ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10519-17-2
Molecular Formula C13H22O2
Molecular Weight (g/mol) 210.32
MDL Number MFCD00036627
InChI Key WVNHQHYIPXYFQX-UHFFFAOYSA-N
Synonym 2-Undecynoic Acid Ethyl Ester
PubChem CID 82694
IUPAC Name ethyl undec-2-ynoate
SMILES CCCCCCCCC#CC(=O)OCC

Specifications

Color Colorless
Boiling Point 128°C
Quantity 25 mL
Formula Weight 210.32
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Undecynoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.