Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-(2-thienyl)-1H-pyrazole-5-carboxylate, 97%, Thermo Scientific™

Catalog No. SPB04031EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SPB04031EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPB04031EA Supplier Thermo Scientific Chemicals Supplier No. SPB04031EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 121195-03-7
Molecular Formula C10H10N2O2S
Molecular Weight (g/mol) 222.262
MDL Number MFCD07772880
InChI Key NTCRDZSJYGZRIE-UHFFFAOYSA-N
Synonym ethyl 5-thien-2-yl-1h-pyrazole-3-carboxylate, ethyl 3-thiophen-2-yl-1h-pyrazole-5-carboxylate, ethyl 5-2-thienyl pyrazole-3-carboxylate, ethyl 5-thiophen-2-yl-1h-pyrazole-3-carboxylate, ethyl 3-2-thienyl-1h-pyrazole-5-carboxylate, ethyl 5-thiophen-2-yl-2h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid, 5-2-thienyl-, ethyl ester, 1h-pyrazole-3-carboxylicacid, 5-2-thienyl-, ethyl ester, acmc-20mpcx, maybridge1_008811
PubChem CID 2736478
IUPAC Name ethyl 5-thiophen-2-yl-1H-pyrazole-3-carboxylate
SMILES CCOC(=O)C1=NNC(=C1)C2=CC=CS2

Specifications

CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 222.27
Percent Purity 95%
Chemical Name or Material Ethyl 5-thien-2-yl-1H-pyrazole-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.