Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 3-[(3-methoxynaphthalen-1-yl)amino]benzoate | 2374831-10-2 | 98.1% | 321.37 | C20H19NO3 | 100MG
SDP

Catalog No. 5000235955
Encompass_Preferred

CDC25B-IN-1 is a small-molecule inhibitor of the CDC25B phosphatase used for biochemical and cellular research. Reported as compound 4a, it inhibits CDC25B with a Ki of 8.5 μM, reduces cell proliferation and colony formation, and induces G2/M cell-cycle arrest. The material is typically supplied as a light brown oil or as a DMSO solution for assay use.

  • Inhibits CDC25B with a reported Ki of 8.5 μM.
  • Reduces cell proliferation and colony formation in cellular assays.
  • Induces G2/M cell-cycle arrest.
  • Molecular formula C20H19NO3; molecular weight 321.37.
  • High purity (approximately 98.1%; may vary by batch).
  • Available in multiple quantities and solution formats for assay convenience.
  • Storage recommendations include refrigeration under inert atmosphere and low-temperature storage in solvent.

Catalog No. 50-002-35955 Supplier Medchemexpress LLC Supplier No. HY126246100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CDC25B-IN-1 is a small-molecule inhibitor of the CDC25B phosphatase used for biochemical and cellular research. Reported as compound 4a, it inhibits CDC25B with a Ki of 8.5 μM, reduces cell proliferation and colony formation, and induces G2/M cell-cycle arrest. The material is typically supplied as a light brown oil or as a DMSO solution for assay use.

  • Inhibits CDC25B with a reported Ki of 8.5 μM.
  • Reduces cell proliferation and colony formation in cellular assays.
  • Induces G2/M cell-cycle arrest.
  • Molecular formula C20H19NO3; molecular weight 321.37.
  • High purity (approximately 98.1%; may vary by batch).
  • Available in multiple quantities and solution formats for assay convenience.
  • Storage recommendations include refrigeration under inert atmosphere and low-temperature storage in solvent.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.