Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3,4-Diaminobenzoate 98.0+%, TCI America™
SDP

Catalog No. E09305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E09305G Supplier TCI America Supplier No. E09305G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37466-90-3
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.207
MDL Number MFCD00179337
InChI Key NUJBTXFFJUGENN-UHFFFAOYSA-N
Synonym 3,4-diaminobenzoic acid ethyl ester, 3,4-diamino-benzoic acid ethyl ester, benzoic acid, 3,4-diamino-, ethyl ester, ethyl3,4-diaminobenzoate, acmc-1afrr, maybridge3_004764, 3,4-diaminobenzoic acid, ethyl ester, 4-ethoxycarbonyl benzene-1,2-diamine, 4-ethoxycarbonyl phenylene-1,2-diamine
PubChem CID 458855
IUPAC Name ethyl 3,4-diaminobenzoate
SMILES CCOC(=O)C1=CC(=C(C=C1)N)N

Specifications

Melting Point 112°C
Color Yellow
Quantity 5 g
Formula Weight 180.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3,4-Diaminobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.