Learn More
Apexbio Technology LLC Ethyl 3,4-dihydroxybenzoate 3943-89-3 1g
SDP
Catalog No. 5000094014
Encompass_Preferred
Ethyl 3 4-dihydroxybenzoate (CAS 3943-89-3) also known as ethyl protocatechuate is a phenolic compound derived from natural sources exhibiting notable antioxidant properties in plant systems It has been shown to activate nitric oxide synthase and enhance mitochondrial ROS production contributing to cardioprotective effects Additionally ethyl 3 4-dihydroxybenzoate inhibits prolyl hydroxylase activity modulating collagen synthesis and demonstrating protective actions on bone tissue In oncology research it induces autophagy and apoptosis in esophageal squamous cell carcinoma cells highlighting its value for studies in cardioprotection bone metabolism and cancer cell regulation
Catalog No. 50-000-94014 Supplier Apexbio Technology LLC Supplier No. A47911000
Ethyl 3 4-dihydroxybenzoate (CAS 3943-89-3) also known as ethyl protocatechuate is a phenolic compound derived from natural sources exhibiting notable antioxidant properties in plant systems It has been shown to activate nitric oxide synthase and enhance mitochondrial ROS production contributing to cardioprotective effects Additionally ethyl 3 4-dihydroxybenzoate inhibits prolyl hydroxylase activity modulating collagen synthesis and demonstrating protective actions on bone tissue In oncology research it induces autophagy and apoptosis in esophageal squamous cell carcinoma cells highlighting its value for studies in cardioprotection bone metabolism and cancer cell regulation