Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3,4-dihydroxybenzoate, 97%

Catalog No. AC325810050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC325810050 5 g Glass bottle
AC325810250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC325810050 Supplier Thermo Scientific Chemicals Supplier No. 325810050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 3943-89-3 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3943-89-3
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002199
InChI Key KBPUBCVJHFXPOC-UHFFFAOYSA-N
Synonym ethyl protocatechuate, 3,4-dihydroxybenzoic acid ethyl ester, ethyl-3,4-dihydroxybenzoate, unii-4ygj96wtbg, benzoic acid, 3,4-dihydroxy-, ethyl ester, edhb, protocatechuic acid ethyl ester, 4ygj96wtbg, 3,4-dihydroxy-benzoic acid ethyl ester
PubChem CID 77547
IUPAC Name ethyl 3,4-dihydroxybenzoate
SMILES CCOC(=O)C1=CC=C(O)C(O)=C1

Specifications

Melting Point 131.0°C to 136.0°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 182.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3, 4-dihydroxybenzoate, 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-529-0

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.