Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3,4-dimethoxyphenylacetate, 98%, Thermo Scientific™

Catalog No. AAA1379630
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1379630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1379630 Supplier Thermo Scientific Chemicals Supplier No. A1379630

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18066-68-7
Molecular Formula C12H16O4
Molecular Weight (g/mol) 224.256
MDL Number MFCD00017272
InChI Key WZKCZNJTDZCNMH-UHFFFAOYSA-N
Synonym ethyl 2-3,4-dimethoxyphenyl acetate, ethyl 3,4-dimethoxyphenylacetate, homoveratric acid, ethyl ester, ethyl dimethoxyphenylacetate, benzeneacetic acid, 3,4-dimethoxy-, ethyl ester, 3,4-dimethoxyphenylacetic acid ethyl ester, ethyl 3,4-dimethoxyphenyl acetate, acetic acid, 3,4-dimethoxyphenyl-, ethyl ester, ethyl homoveratrate
PubChem CID 87441
IUPAC Name ethyl 2-(3,4-dimethoxyphenyl)acetate
SMILES CCOC(=O)CC1=CC(=C(C=C1)OC)OC

Specifications

Boiling Point 159°C to 160°C (4 mmHg)
Quantity 250 g
Beilstein 2697968
Formula Weight 224.26
Percent Purity 98%
Chemical Name or Material Ethyl 3,4-dimethoxyphenylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.