Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 3-(4-fluorophenyl)-1H-pyrazole-5-carboxylate

Catalog No. JRD00281G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JRD00281G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JRD00281G Supplier Sigma Aldrich Supplier No. JRD00281G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Building block used as a precursor to the synthesis of azaspiro[3.4]octanes developed in the carreira group
  • This substrate can be readily functionalized via enolization at the 3-position in the presence of lda

Specifications

Molecular Formula C12H11FN2O2
Linear Formula C12H11FN2O2
MDL Number MFCD08445938
Quantity 5 g
Molecular Weight (g/mol) 234.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.