Learn More
Medchemexpress LLC Ethyl 3-(4-fluorophenyl)-3-oxopropanoate | 1999-00-4 | ≥97.0% | C11H11FO3 | 10 G
SDP
Catalog No. 5000434973
Encompass_Preferred
Ethyl 3-(4-fluorophenyl)-3-oxopropanoate is a biochemical reagent suitable for life science related research. It appears as a colorless to light yellow liquid with a density of 1.174 g/cm3. This product is for research use only and not intended for medical applications.
  • Can be used as a biological material or organic compound
  • Colorless to light yellow liquid appearance
  • Density: 1.174 g/cm3
Catalog No. 50-004-34973 Supplier Medchemexpress LLC Supplier No. HY7716010G
Ethyl 3-(4-fluorophenyl)-3-oxopropanoate is a biochemical reagent suitable for life science related research. It appears as a colorless to light yellow liquid with a density of 1.174 g/cm3. This product is for research use only and not intended for medical applications.
  • Can be used as a biological material or organic compound
  • Colorless to light yellow liquid appearance
  • Density: 1.174 g/cm3