Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3,5-bis(trifluoromethyl)benzoate, 97%, Thermo Scientific™

Catalog No. AC326360050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC326360050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC326360050 Supplier Thermo Scientific Chemicals Supplier No. 326360050
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 96617-71-9
Molecular Formula C11H8F6O2
Molecular Weight (g/mol) 286.17
MDL Number MFCD01320684
InChI Key NBWZJMOEFTYMOF-UHFFFAOYSA-N
Synonym ethyl 3,5-bis trifluoromethyl benzoate, 3,5-bis-trifluoromethyl-benzoic acid ethyl ester, 3,5-bis trifluoromethyl benzoic acid ethyl ester, rarechem al bi 0207, acmc-20al07, ethyl 3?5-bis trifluoromethyl benzoate, ethyl 3 5-bis trifluoromethyl benzoate, benzoic acid, 3,5-bis trifluoromethyl-, ethyl ester
PubChem CID 2733458
IUPAC Name ethyl 3,5-bis(trifluoromethyl)benzoate
SMILES CCOC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4145 to 1.4165
Quantity 5 g
Formula Weight 286.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 3, 5-bis(trifluoromethyl)benzoate, 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.