Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3,5-dimethyl-1H-4-pyrazolecarboxylate, 97%, Thermo Scientific™

Catalog No. CC01223DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC01223DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC01223DA Supplier Thermo Scientific Chemicals Supplier No. CC01223DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 35691-93-1
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.196
MDL Number MFCD00159638
InChI Key BCKARVLFIJPHQU-UHFFFAOYSA-N
Synonym ethyl 3,5-dimethyl-1h-4-pyrazolecarboxylate, ethyl 3,5-dimethylpyrazole-4-carboxylate, 3,5-dimethylpyrazole-4-carboxylic acid ethyl ester, 3,5-dimethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 4-pyrazolecarboxylic acid, 3,5-dimethyl-, ethyl ester, 3.5. dimethylpyrazol 4-carbonsaeureaethyl ester german, 3.5. dimethylpyrazol 4-carbonsaeureaethyl ester, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrazole, 1h-pyrazole-4-carboxylic acid, 3,5-dimethyl-, ethyl ester, dee
PubChem CID 215436
IUPAC Name ethyl 3,5-dimethyl-1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(NN=C1C)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 168.19
Percent Purity 97%
Chemical Name or Material Ethyl 3, 5-dimethyl-1H-4-pyrazolecarboxylate
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.