Learn More
Apexbio Technology LLC ETHYL 3-AMINOBENZOATE MS-50g
SDP
Catalog No. 5000897630
Encompass_Preferred
Ethyl 3-Aminobenzoate (methanesulfonate) (CAS No 886-86-2) is commonly used in biomedical research particularly in the study of pain mechanisms and local anesthetics Originating from its role as a derivative of benzoic acid it primarily acts by inhibiting voltage-gated sodium channels which modulates nerve signal transmission Its in vitro activity typically ranges from nanomolar to low micromolar concentrations This compound is frequently applied in various experimental contexts including cell models and animal studies to investigate nerve impulse propagation and potential therapeutic uses in pain management It is also utilized in drug screening assays to evaluate its efficacy and selectivity in modulating sodium channel activity Experimental concentrations typically depend on the specific research design ensuring alignment with the desired experimental outcomes
Catalog No. 50-008-97630 Supplier Apexbio Technology LLC Supplier No. C361950000
Add to Cart
Add to Cart
Ethyl 3-Aminobenzoate (methanesulfonate) (CAS No 886-86-2) is commonly used in biomedical research particularly in the study of pain mechanisms and local anesthetics Originating from its role as a derivative of benzoic acid it primarily acts by inhibiting voltage-gated sodium channels which modulates nerve signal transmission Its in vitro activity typically ranges from nanomolar to low micromolar concentrations This compound is frequently applied in various experimental contexts including cell models and animal studies to investigate nerve impulse propagation and potential therapeutic uses in pain management It is also utilized in drug screening assays to evaluate its efficacy and selectivity in modulating sodium channel activity Experimental concentrations typically depend on the specific research design ensuring alignment with the desired experimental outcomes