Learn More
Medchemexpress LLC Ethyl 3-azidopropanoate | 40139-55-7 | MFCD24452394 | >95.0% | 143.14 g·mol⁻1 | C5H9N3O2 | 25 MG
SDP
Catalog No. 5000284004
Encompass_Preferred
Ethyl 3-azidopropanoate is an ethyl ester bearing a terminal azide group used as a building block in organic synthesis and click-chemistry applications. The azide functionality readily undergoes cycloaddition with alkynes to form stable triazole linkages, enabling modular assembly and bioconjugation.
  • Azide-functional ethyl ester for click chemistry.
  • Reacts in copper-catalyzed and strain-promoted cycloadditions to yield triazoles.
  • High purity (≥95.0%) suitable for synthetic applications.
  • Liquid, colorless to light yellow, facilitating handling and dosing.
  • Recommended storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Supplied as a 25 mg aliquot for small-scale synthesis and screening.
Catalog No. 50-002-84004 Supplier Medchemexpress LLC Supplier No. HYW59819825MG
Add to Cart
Add to Cart
Ethyl 3-azidopropanoate is an ethyl ester bearing a terminal azide group used as a building block in organic synthesis and click-chemistry applications. The azide functionality readily undergoes cycloaddition with alkynes to form stable triazole linkages, enabling modular assembly and bioconjugation.
  • Azide-functional ethyl ester for click chemistry.
  • Reacts in copper-catalyzed and strain-promoted cycloadditions to yield triazoles.
  • High purity (≥95.0%) suitable for synthetic applications.
  • Liquid, colorless to light yellow, facilitating handling and dosing.
  • Recommended storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Supplied as a 25 mg aliquot for small-scale synthesis and screening.