Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-(Benzylamino)propionate 95.0+%, TCI America™
SDP

Catalog No. B1579500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1579500ML 500 mL
B157925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1579500ML Supplier TCI America Supplier No. B1579500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 23583-21-3 | C12H17NO2 | 207.273 g/mol

Chemical Identifiers

CAS 23583-21-3
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD00085541
InChI Key HCTJHQFFNDLDPF-UHFFFAOYSA-N
Synonym ethyl 3-benzylamino propanoate, ethyl 3-benzylamino propionate, ethyl n-benzyl-beta-alaninate, n-benzyl-beta-alanine ethyl ester, 3-benzylamino-propionic acid ethyl ester, 3-benzylaminopropionic acid ethyl ester, n-benzyl-3-aminopropionic acid ethyl ester, ethyl n-benzyl-, a-alaninate, ethyl3-benzylamino propanoate
PubChem CID 90196
IUPAC Name ethyl 3-(benzylamino)propanoate
SMILES CCOC(=O)CCNCC1=CC=CC=C1

Specifications

Boiling Point 135°C
Color Yellow
Quantity 500 mL
Formula Weight 207.27
Percent Purity ≥95.0% (GC)
Grade Laboratory
Physical Form Liquid
Chemical Name or Material Ethyl 3-(Benzylamino)propionate
EINECSNumber (3)-4218
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.