Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-Chloropropionate 97.0+%, TCI America™
SDP

Catalog No. C127425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C127425G 25 g
C1274100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C127425G Supplier TCI America Supplier No. C127425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 623-71-2
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00000989
InChI Key ZCLGVXACCAZJOX-UHFFFAOYSA-N
Synonym ethyl 3-chloropropionate, 3-chloropropionic acid ethyl ester, propanoic acid, 3-chloro-, ethyl ester, ethyl beta-chloropropionate, propionic acid, 3-chloro-, ethyl ester, ethyl .beta.-chloropropionate, 3-chloropropionicacidethylester, 3-chloro-propanoicacidethylester, ethyl-3-chlorpropanoat, ethyl3-chloropropionate
PubChem CID 69341
IUPAC Name ethyl 3-chloropropanoate
SMILES CCOC(=O)CCCl

Specifications

Color Colorless
Boiling Point 162°C
Quantity 25 g
UN Number 3272
Formula Weight 136.58
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Chloropropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.