Learn More
Medchemexpress LLC Ethyl 3-coumarincarboxylate | 1846-76-0 | MFCD00016964 | 99.8% | 218.21 g/mol | C12H10O4 | 5 G
SDP
Catalog No. 5000302767
Encompass_Preferred
Ethyl 3-coumarincarboxylate (ethyl 2-oxo-2H-chromene-3-carboxylate) is a coumarin derivative used as a research reagent and synthetic intermediate. It is commonly employed as a pseudo-template in molecularly imprinted polymer synthesis and in the preparation of coumarin-based compounds. Typical properties include formula C12H10O4, molecular weight 218.21 g/mol, and reported purity ~99.8%.
  • High reported purity for reliable experimental results.
  • Useful intermediate for synthesis of coumarin derivatives.
  • Functions as a pseudo-template for molecularly imprinted polymer (MIP) preparation.
  • Available in small laboratory pack sizes for bench-scale work.
  • White solid with documented physicochemical data for reproducibility.
Catalog No. 50-003-02767 Supplier Medchemexpress LLC Supplier No. HYW0140815G
Ethyl 3-coumarincarboxylate (ethyl 2-oxo-2H-chromene-3-carboxylate) is a coumarin derivative used as a research reagent and synthetic intermediate. It is commonly employed as a pseudo-template in molecularly imprinted polymer synthesis and in the preparation of coumarin-based compounds. Typical properties include formula C12H10O4, molecular weight 218.21 g/mol, and reported purity ~99.8%.
  • High reported purity for reliable experimental results.
  • Useful intermediate for synthesis of coumarin derivatives.
  • Functions as a pseudo-template for molecularly imprinted polymer (MIP) preparation.
  • Available in small laboratory pack sizes for bench-scale work.
  • White solid with documented physicochemical data for reproducibility.