Learn More
Medchemexpress LLC Ethyl 3-hydroxybutyrate | 5405-41-4 | 99.99% | 132.16 | 1 ML
SDP
Catalog No. 5000454965
Encompass_Preferred
Ethyl 3-hydroxybutyrate is a fragrance that is naturally found in wine and in *Tribolium castaneum*. It has been shown to increase 3-HB levels in serum, promote the TCA cycle and protein synthesis, and improve metabolic homeostasis. Additionally, it reduces inflammation and enhances antioxidant capacity, which can alleviate cancer cachexia-associated skeletal muscle atrophy in CT26 xenograft mouse models.
  • Fragrance compound
  • Increases 3-HB levels in serum
  • Promotes TCA cycle and protein synthesis
  • Improves metabolic homeostasis
  • Reduces inflammation responses
  • Enhances antioxidant capacity
  • Alleviates cancer cachexia-associated skeletal muscle atrophy in CT26 xenograft mouse models
Catalog No. 50-004-54965 Supplier Medchemexpress LLC Supplier No. HYW01270110MM/1ML
Ethyl 3-hydroxybutyrate is a fragrance that is naturally found in wine and in *Tribolium castaneum*. It has been shown to increase 3-HB levels in serum, promote the TCA cycle and protein synthesis, and improve metabolic homeostasis. Additionally, it reduces inflammation and enhances antioxidant capacity, which can alleviate cancer cachexia-associated skeletal muscle atrophy in CT26 xenograft mouse models.
  • Fragrance compound
  • Increases 3-HB levels in serum
  • Promotes TCA cycle and protein synthesis
  • Improves metabolic homeostasis
  • Reduces inflammation responses
  • Enhances antioxidant capacity
  • Alleviates cancer cachexia-associated skeletal muscle atrophy in CT26 xenograft mouse models