Learn More
Medchemexpress LLC Ethyl 3-hydroxybutyrate | 5405-41-4 | 99.99% | 132.16 | 100 G
SDP
Catalog No. 5000454819
Encompass_Preferred
Ethyl 3-hydroxybutyrate is a fragrance found in wine and the insect *Tribolium castaneum*. It has been studied for its therapeutic potential in alleviating skeletal muscle wasting in cancer cachexia by increasing 3-HB levels and promoting the TCA cycle.
  • Increases 3-HB levels in serum
  • Promotes TCA cycle and protein synthesis
  • Improves metabolic homeostasis
  • Reduces inflammation and enhances antioxidant capacity
  • Alleviates cancer cachexia-associated skeletal muscle atrophy
Catalog No. 50-004-54819 Supplier Medchemexpress LLC Supplier No. HYW012701100G
Add to Cart
Add to Cart
Ethyl 3-hydroxybutyrate is a fragrance found in wine and the insect *Tribolium castaneum*. It has been studied for its therapeutic potential in alleviating skeletal muscle wasting in cancer cachexia by increasing 3-HB levels and promoting the TCA cycle.
  • Increases 3-HB levels in serum
  • Promotes TCA cycle and protein synthesis
  • Improves metabolic homeostasis
  • Reduces inflammation and enhances antioxidant capacity
  • Alleviates cancer cachexia-associated skeletal muscle atrophy