Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethyl 3 hydroxybutyrate 97
Catalog No. 502914363
Encompass
Ethyl 3-hydroxybutyrate is one of the main aroma components of wine. It occurs naturally in fruits such as pineapple tamarillo fruit caja fruit and naranjilla fruit.
Catalog No. 50-291-4363 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W34280710KGK
Ethyl 3-hydroxybutyrate is one of the main aroma components of wine. It occurs naturally in fruits such as pineapple tamarillo fruit caja fruit and naranjilla fruit.