Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-mercaptopropionate, 98%

Catalog No. AC137412500
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC137412500 250 g Glass bottle
AC137410050 5 g Glass bottle
AC137410250 25 g Glass bottle
AC137411000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC137412500 Supplier Thermo Scientific Chemicals Supplier No. 137412500

CAS: 5466-06-8 | ClH4NO | 69.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5466-06-8
IUPAC Name ethyl 3-sulfanylpropanoate

Specifications

Color Colorless to Yellow
Density 1.0590g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H10O2S
Refractive Index 1.4560 to 1.4580 (20°C, 589nm)
Linear Formula HSCH2CH2CO2C2H5
Quantity 250 g
Synonym ethyl 3-mercaptopropionate, propanoic acid, 3-mercapto-, ethyl ester, 3-mercaptopropanoic acid ethyl ester, 3-mercaptopropionic acid ethyl ester, ethyl 3-mercaptopropanoate, ethyl beta-mercaptopropionate, unii-22gqz8p70d, propionic acid, 3-mercapto-, ethyl ester, ethyl 3-mercaptopropanoic acid, ethyl .beta.-mercaptopropionate
InChI Key CJQWLNNCQIHKHP-UHFFFAOYSA-N
SMILES CCOC(=O)CCS
Molecular Weight (g/mol) 134.19
PubChem CID 21625
Formula Weight 134.2
Percent Purity ≥97.5% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-mercaptopropionate, 99%
Show More Show Less
Hazard Category Warning
EINECSNumber 226-771-5
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.