Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-mercaptopropionate, 98%

Catalog No. p-130821
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC137410050 5 g Glass bottle
AC137410250 25 g Glass bottle
AC137411000 100 g Glass bottle
AC137412500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AC137410050 Supplier Thermo Scientific Chemicals Supplier No. 137410050
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 5466-06-8 | ClH4NO | 69.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5466-06-8
MDL Number MFCD00051089
ChEBI CHEBI:5807
IUPAC Name ethyl 3-sulfanylpropanoate

Specifications

Color Colorless to Yellow
Density 1.0590g/mL
Boiling Point 75.0°C to 76.0°C (10.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula ClH4NO
Refractive Index 1.4560 to 1.4580
Linear Formula HSCH2CH2CO2C2H5
Quantity 5 g
Beilstein 03,555
Specific Gravity 1.059
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
SMILES Cl.NO
Molecular Weight (g/mol) 69.49
PubChem CID 443297
Formula Weight 134.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 3-mercaptopropionate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 226-771-5
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.