Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-Mercaptopropionate, Approx. 99%, Spectrum™ Chemical
SDP

Catalog No. 18602492 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-492 500 g Amber Glass Bottle
18-602-490 25 g Amber Glass Bottle
18-602-491 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-492 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E2745500GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

E2745, 5466-06-8, C5H10O2S

Spectrum™ Chemical Ethyl 3-Mercaptopropionate

Chemical Identifiers

CAS 5466-06-8
Molecular Formula C5H10O2S
Molecular Weight (g/mol) 134.19
InChI Key CJQWLNNCQIHKHP-UHFFFAOYSA-N
IUPAC Name ethyl 3-sulfanylpropanoate
SMILES CCOC(=O)CCS

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity ∼99%
Grade Reagent
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.