Learn More
Medchemexpress LLC Ethyl 3-oxohexanoate | 3249-68-1 | MFCD00009401 | 96.0% | 158.20 g/mol | C8H14O3 | 100 G
SDP
Catalog No. 5000303636
Encompass_Preferred
Ethyl butyrylacetate (ethyl 3-oxohexanoate) is an organic beta-keto ester (C8H14O3, MW 158.20 g/mol) used as a flavor and fragrance ingredient and as a synthetic building block in organic synthesis. It has a fruity aroma and is supplied for research use; applications include flavor formulation, perfumery, and laboratory-scale chemical synthesis.
  • Fruity aroma useful for flavor and fragrance formulation.
  • Useful synthetic intermediate in organic reactions and building-block chemistry.
  • Available in multiple sizes suitable for laboratory use.
  • Purity typically around 96.0% for reagent-grade applications.
  • Supplied for research use only.
Catalog No. 50-003-03636 Supplier Medchemexpress LLC Supplier No. HYW015487100G
Ethyl butyrylacetate (ethyl 3-oxohexanoate) is an organic beta-keto ester (C8H14O3, MW 158.20 g/mol) used as a flavor and fragrance ingredient and as a synthetic building block in organic synthesis. It has a fruity aroma and is supplied for research use; applications include flavor formulation, perfumery, and laboratory-scale chemical synthesis.
  • Fruity aroma useful for flavor and fragrance formulation.
  • Useful synthetic intermediate in organic reactions and building-block chemistry.
  • Available in multiple sizes suitable for laboratory use.
  • Purity typically around 96.0% for reagent-grade applications.
  • Supplied for research use only.