Learn More
Medchemexpress LLC Ethyl 3-[(tert-butylcarbamoyl)(N-[(4-chlorophenyl)methyl]formamido)methyl]-6-chloro-1H-indole-2-carboxylate | 1364488-67-4 | 98.3% | 50 MG
SDP
Catalog No. 5000232395
Encompass_Preferred
YH239-EE is the ethyl ester of a small-molecule MDM2/p53 antagonist used in vitro to disrupt the p53-MDM2 interaction, inducing cell-cycle arrest and apoptosis in p53 wild-type acute myeloid leukemia (AML) cell lines. Supplied as a high-purity solid and available in multiple sizes and solution formats for cellular and biochemical assays.
  • Antagonizes the p53-MDM2 interaction in vitro.
  • Induces cell-cycle arrest and apoptosis in p53 wild-type AML cells.
  • High purity (98.3%) solid, white to off-white, suitable for research assays.
  • Available in multiple quantities and as a 10 mM solution in DMSO for cell-based studies.
  • Molecular weight 504.41 g/mol; formula C25H27Cl2N3O4.
Catalog No. 50-002-32395 Supplier Medchemexpress LLC Supplier No. HY1228750MG
YH239-EE is the ethyl ester of a small-molecule MDM2/p53 antagonist used in vitro to disrupt the p53-MDM2 interaction, inducing cell-cycle arrest and apoptosis in p53 wild-type acute myeloid leukemia (AML) cell lines. Supplied as a high-purity solid and available in multiple sizes and solution formats for cellular and biochemical assays.
  • Antagonizes the p53-MDM2 interaction in vitro.
  • Induces cell-cycle arrest and apoptosis in p53 wild-type AML cells.
  • High purity (98.3%) solid, white to off-white, suitable for research assays.
  • Available in multiple quantities and as a 10 mM solution in DMSO for cell-based studies.
  • Molecular weight 504.41 g/mol; formula C25H27Cl2N3O4.