Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-(trifluoromethyl)crotonate, (E)+(Z), 96%

Catalog No. AAL0968809
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0968809 10 g
AAL0968804 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0968809 Supplier Thermo Scientific Chemicals Supplier No. L0968809
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 24490-03-7 | C7H9F3O2 | 182.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24490-03-7
Molecular Formula C7H9F3O2
Molecular Weight (g/mol) 182.14
MDL Number MFCD00040846
InChI Key OSZLARYVWBUKTG-SNAWJCMRSA-N
Synonym ethyl 4,4,4-trifluoro-3-methylbut-2-enoate, ethyl 3-trifluoromethyl crotonate, ethyl 2e-4,4,4-trifluoro-3-methylbut-2-enoate, ethyl 3-trifluoromethyl crotonate, cis + trans, ethyl e-4,4,4-trifluoro-3-methylbut-2-enoate, e-3-trifluoromethyl-2-butenoic acid ethyl ester
PubChem CID 5838607
IUPAC Name ethyl (E)-4,4,4-trifluoro-3-methylbut-2-enoate
SMILES CCOC(=O)\C=C(/C)C(F)(F)F

Specifications

Boiling Point 129°C to 130°C
Flash Point 32°C (89°F)
Refractive Index 1.378
Quantity 10 g
UN Number UN3272
Beilstein 1910081
Formula Weight 182.14
Percent Purity 96%
Chemical Name or Material Ethyl 3-(trifluoromethyl)crotonate, (E)+(Z)
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.