Learn More
eMolecules​ 10601-80-6 | Ethyl 3,3-diethoxypropionate | Ambeed | MFCD00009865 | 190.239 | C9H18O4 | 97.000 | CCOC(CC(=O)OCC)OCC | 100g | 552648994
Catalog No. 502695772 Shop All eMolecules Inc. Products
Encompass_Preferred
Ethyl 3,3-diethoxypropionate | Ambeed | 10601-80-6 | MFCD00009865 | 190.239 | C9H18O4 | 97.000 | CCOC(CC(=O)OCC)OCC | 100g | 552648994
Catalog No. 50-269-5772 Supplier eMolecules​ Supplier No. 552648994
Ethyl 3,3-diethoxypropionate | Ambeed | 10601-80-6 | MFCD00009865 | 190.239 | C9H18O4 | 97.000 | CCOC(CC(=O)OCC)OCC | 100g | 552648994