Learn More
Medchemexpress LLC Ethyl 4-((1-hydroxy-2-phenyl-1H-indol-3-yl)(pyridin-2-yl)methyl)piperazine-1-carboxylate | 622795-76-0 | 98.0% | 456.54 g/mol | C27H28N4O3 | 10MG
SDP
Catalog No. 5000238578
Encompass_Preferred
PIP-199 is a small-molecule inhibitor that selectively disrupts the RMI (RecQ-mediated genome instability protein) core complex/MM2 protein-protein interaction. It is supplied as a research reagent (CAS 622795-76-0) with molecular formula C27H28N4O3 and is used in studies of DNA repair and in research on sensitizing tumor cells to DNA crosslinking agents.
  • Selective inhibitor of RMI-MM2 protein-protein interaction.
  • Useful for studying DNA repair and tumor sensitization to crosslinking agents.
  • Characterized by a molecular weight of 456.54 g/mol and formula C27H28N4O3.
  • Purity typically 98.0% for research applications.
  • Available in small-scale quantities suitable for laboratory experiments.
Catalog No. 50-002-38578 Supplier Medchemexpress LLC Supplier No. HY12432510MG
PIP-199 is a small-molecule inhibitor that selectively disrupts the RMI (RecQ-mediated genome instability protein) core complex/MM2 protein-protein interaction. It is supplied as a research reagent (CAS 622795-76-0) with molecular formula C27H28N4O3 and is used in studies of DNA repair and in research on sensitizing tumor cells to DNA crosslinking agents.
  • Selective inhibitor of RMI-MM2 protein-protein interaction.
  • Useful for studying DNA repair and tumor sensitization to crosslinking agents.
  • Characterized by a molecular weight of 456.54 g/mol and formula C27H28N4O3.
  • Purity typically 98.0% for research applications.
  • Available in small-scale quantities suitable for laboratory experiments.