Learn More
Medchemexpress LLC Ethyl 4-(3-(4-(1H-pyrazol-1-yl)benzyl)ureido)benzoate | 1005883-72-6 | 98.1% | 364.40 g/mol | C20H20N4O3 | 25MG
SDP
Catalog No. 5000237804
Encompass_Preferred
This small-molecule inhibitor is selective for aquaporin-7 (AQP7) and intended for laboratory research. It inhibits mouse AQP7 with an IC50 of ~0.2 μM and shows weaker activity against AQP3 and AQP9 (IC50s ~0.7 μM and ~1.1 μM). The compound has formula C20H20N4O3, molecular weight 364.40 g/mol, and a reported purity of 98.1%.
  • Potent and selective inhibitor of aquaporin-7 (AQP7).
  • Submicromolar activity against mAQP7; less potent against mAQP3 and mAQP9.
  • High reported purity (98.1%).
  • High solubility in DMSO (≥280 mg/mL) with reported in vivo formulation options.
  • Stable when stored as powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in multiple solid and solution formats for research use.
Catalog No. 50-002-37804 Supplier Medchemexpress LLC Supplier No. HY12607425MG
This small-molecule inhibitor is selective for aquaporin-7 (AQP7) and intended for laboratory research. It inhibits mouse AQP7 with an IC50 of ~0.2 μM and shows weaker activity against AQP3 and AQP9 (IC50s ~0.7 μM and ~1.1 μM). The compound has formula C20H20N4O3, molecular weight 364.40 g/mol, and a reported purity of 98.1%.
  • Potent and selective inhibitor of aquaporin-7 (AQP7).
  • Submicromolar activity against mAQP7; less potent against mAQP3 and mAQP9.
  • High reported purity (98.1%).
  • High solubility in DMSO (≥280 mg/mL) with reported in vivo formulation options.
  • Stable when stored as powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in multiple solid and solution formats for research use.